Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurine, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18614325 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-325 500 g Poly Bottle
18-614-321 100 g Poly Bottle
18-614-320 1 kg Poly Bottle
18-614-323 5 kg Poly Pail
18-614-322 25 kg Fiber Drum
18-614-324 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-614-325 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TA122500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-35-7

Specifications

CAS Max % 1
Loss on Drying 0.001
Packaging Poly Bottle
Quantity 500 g
Molecular Weight (g/mol) 125.14
Percent Purity 0.985
Grade Reagent
Physical Form Crystalline

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.