Learn More
ACROBiosystems Human Fc gamma RIIIA / CD16a (F176, S197P), His Tag (CDA-H52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (F176, S197P) (Accession # P08637-1 (F176, S197P)).
Supplier:  ACROBiosystems CDAH52H6100UG
Encompass_Preferred
CD16 is a low affinity Fc receptor, and has been identified as Fc receptors FcγRIIIa (CD16a) and FcγRIIIb (CD16b). These receptors bind to the Fc portion of IgG antibodies. CD16 encoded by two different highly homologous genes in a cell type-specific manner.CD16 is found on the surface of natural killer cells, neutrophil polymorphonuclear leukocytes, monocytes and macrophages.CD16a antigen is also known as Low affinity immunoglobulin gamma Fc region receptor III-A, Fc-gamma RIII-alpha. CD16b is a low-affinity, GPI-linked receptor expressed by neutrophils and eosinophils, whereas CD16a is an intermediate affinity polypeptide-anchored transmembrane glycoprotein expressed natural killer cells, macrophages, subpopulation of T-cells, immature thymocytes and placentaltrophoblasts.
Catalog No. 50-223-0723