Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIB/CD16b Mouse anti-Human, Clone: 245514, R&D Systems™

Catalog No. MAB1597SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1597SP 25 μg
MAB1597 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1597SP Supplier R&D Systems Supplier No. MAB1597SP
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fc gamma RIIIB/CD16b Monoclonal antibody MAB1597SP Specifically detects Fc gamma RIIIB/CD16b in Human samples. It is validated for Flow Cytometry, Western Blotting

Specifications

Antigen Fc gamma RIIIB/CD16b
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 245514
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O75015
Gene Alias CD16, CD16b, CD16b antigen, Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc fragment of IgG, low affinity IIIb, receptor for (CD16), FCG3, Fc-gamma receptor IIIb (CD 16), Fc-gamma RIII, Fc-gamma RIIIb, Fc-gamma RIII-beta, FCGR3, FCGR3B, FcgRIIIB, FcR-10, fcRIII, fcRIIIb, IGFR3, IgG Fc receptor III-1, low affinity immunoglobulin gamma Fc region receptor III-B
Gene Symbols FCGR3B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIB/CD16b Thr20-Gln208 Accession # O75015
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2215
Test Specificity Detects human Fc gamma RIIIB/CD16b in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Fc gamma RIIA or recombinant mouse Fc gamma RIII is observed. In flow cytometry of whole blood, recognition of Fc gamma RIIIB on the granulocyte population, but not Fc gamma RIIIA on NK cells, is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.