Learn More
Enzo Life Sciences Fc (human) : CD40L (soluble) (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522072C010
Encompass_Preferred
Alternative name: CD154, TNFSF 5, gp39. Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse CD40L (CD154) (aa 115-260) is fused at the N-terminus to the Fc portion of human IgG1 and a linker peptide (20 aa). Specificity: Binds to human and mouse CD40. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9404
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.