Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fc Receptor Binding Inhibitor Polyclonal Antibody, Functional Grade, eBioscience™, Invitrogen™

Catalog No. p-7218349
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-916-138 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-916-138 Supplier Invitrogen™ Supplier No. 16916138
Only null left
Add to Cart
Add to Cart

Polyclonal Antibody

The human FcgammaR-binding inhibitor can be used to inhibit the non-specific Fc-gamma receptor (FcgammaR)-mediated binding of mouse monoclonal antibodies used for flow cytometric analysis of human tissue. Four different classes of Fcgamma receptors have been identified: FcgammaRI (CD64), FcgammaRII (CD32), and FcgammaRIII (CD16), and they are expressed at varying levels in multiple cell lineages including high expression in myeloid, granulocyte and B cells. The biological function of the FcgammaR, including initiation of endocytosis, phagocytosis and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to human FcgammaR varies depending on the isotype of the MAb. Furthermore, different monoclonal antibodies of the same isotype will display different binding to human FcgammaRs.

Four different classes of Fcy receptors have been identified: FcyRI (CD64), FcyRII (CD32), and FcyRIII (CD16), and they are expressed at varying levels in multiple cell lineages including high expression in myeloid, granulocyte and B cells. The biological function of the FcyR, including initiation of endocytosis, phagocytosis and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to human FcyR varies depending on the isotype of the monoclonal antibody. Furthermore, different monoclonal antibodies of the same isotype will display different binding to human FcyRs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc Receptor Binding Inhibitor
Applications Control, Flow Cytometry, Functional Assay
Classification Polyclonal
Concentration 20 μL/Test
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Alias Fc blocking; FcR BI; FcR Inhibitor
Host Species Not Available
Purification Method Affinity chromatography
Quantity 5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.