Learn More
Julabo USA Inc Each, FC1600S recirculating chiller 1.55 kW
SDP
Supplier:  Julabo USA Inc 9600163.13
Encompass_Preferred
Chillers, Julabo, FC1600S Recirculating Cooler -15...+80 °C, 1.55kW cooling power; pump 22L/min to 17.4 PSI. JULABO FC units feature two separate Multi-Displays (LED) for actual and set point values. Models designated FCW are water-cooled which reduces the noise level and reduced heat generation to the laboratory environment. Units are conveniently filled from the front and have a removable venting grid.FC units feature a built-in 1.2kW heater to facilitate cooling and heating applications for temperatures from -25 °C to +80 °C with an adjustable ratio for inlet/outlet temperature. Models FC1600T and FCW2500T have a Pt100 sensor connection for external measurement and control, temperature recorder and connections for external programming. All FC units have RS232 interface with connections for standby input and alarm output. FC models supplied with 2 barbed fittings for 8 and 12 mm internal ID tubing. Pump connections M16x1 male. 230V/60Hz.
Catalog No. 50-753-3971
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov