Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FCAR/CD89 Antibody Pair [HRP]
SDP

Catalog No. NB009952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009952 5 Plates
NB009951 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB009952 Supplier Novus Biologicals™ Supplier No. NBP2794565PLATES
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 7.8125 pg/mL
Assay Sensitivity 7.81-500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human FCAR/CD89.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Gene ID (Entrez) 2204
Gene Symbol FCAR
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 30 ng of recombinant FCAR/CD89.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-FCAR/CD89 monoclonal antibody,
  • Detection Antibody: 0.25 mg/mL rabbit anti-FCAR/CD89 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 5 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target FCAR/CD89
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1.0 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.