Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCAR (soluble) Monoclonal Antibody (MIP7c)

Catalog No. MA172500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA172500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA172500 Supplier Invitrogen™ Supplier No. MA172500

Mouse Monoclonal Antibody

This antibody recognizes the EC1 domain of the FcaRI extracellular region.

This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCAR (soluble)
Applications Flow Cytometry, Inhibition Assays, Radioimmune Assays (RIA)
Classification Monoclonal
Clone MIP7c
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Fcar
Gene Accession No. P24071
Gene Alias CD89; CTB-61M7.2; Fc alpha receptor; Fc fragment of IgA receptor; Fc fragment of IgA, receptor for; FcalphaRI; FCAR; IgA Fc receptor; immunoglobulin A Fc receptor; immunoglobulin A Fc receptor transcript variant 1; immunoglobulin A Fc receptor transcript variant 3; immunoglobulin A Fc receptor transcript variant 7; immunoglobulin A Fc receptor transcript variant 9; immunoglobulin A Fc receptor transcript variant deltaEC1; immunoglobulin A Fc receptor transcript variant deltaEC1 delta TM/C; immunoglobulin alpha Fc receptor
Gene Symbols Fcar
Host Species Mouse
Immunogen Recombinant soluble CD89.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2204
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.