Learn More
Apexbio Technology LLC FCCP(Synonyms: Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone), 10mg, CAS: 370-86-5.
SDP
Supplier:  Apexbio Technology LLC B500410
Encompass_Preferred
FCCP (CAS 370-86-5) chemically known as carbonyl cyanide p-trifluoromethoxyphenylhydrazone is a lipophilic mitochondrial uncoupler frequently used in cellular biology research It functions by transporting protons across mitochondrial membranes dissipating the proton gradient essential for ATP synthesis FCCP exhibits potent inhibitory activity (IC 0 51 M in T47D cells) effectively disrupting oxidative phosphorylation Experimentally FCCP increases cellular oxygen consumption and suppresses hypoxia-inducible factors (HIF-1 and HIF-2 ) reducing downstream gene expression such as VEGF In rodent embryos FCCP impairs mitochondrial function resulting in decreased ATP production lower birth weight and altered metabolic phenotypes in offspring highlighting its utility in studying metabolic regulation and mitochondrial biology
Catalog No. 50-000-59996
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.