Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595887 Supplier Invitrogen™ Supplier No. PA595887
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCITDCVMAE IEKLGQKYRV ALRIAKDPRF ERLPCTHKGT YADDCLVQRV TQHKCYIVAT VDRDLKRRIR KIPGVPIMYI SNHRYNIERM PDDYGAPRF.

Essential protein involved in pre-rRNA processing and 40S ribosomal subunit assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FCF1
Gene Accession No. Q9Y324
Gene Alias Bka; C14orf111; CGI-35; FCF1; FCF1 rRNA-processing protein; FCF1 small subunit; RGD1310899; rRNA-processing protein FCF1 homolog; UTP24
Gene Symbols FCF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-198 of human FCF1 (NP_0570461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299198, 51077
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.