Learn More
Promega FcgammaRIIa-H ADCP Reporter Bioassay, Core Kit, 1 each
Catalog No. PRG9991 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRG9991 1 Ea.
1 options
Catalog No. PRG9991 Supplier Promega Supplier No. G9991
The FcgammaRIIa-H ADCP Reporter Bioassay is a biologically relevant MOA-based assay that can be used to measure the potency and stability of antibodies and other biologics that specifically bind and activate FcgammaRIIa.
  • Measure Potency and Stability of Biologics that Bind and Activate ADCP through FcγRIIa−H
  • Available in two kit formats:
  • Complete, everything you need to get started
  • Core, used with customer−defined Ab and target cells
Specifications
Quantity 1 Ea.