Learn More
Promega FcgammaRIIa-H ADCP Reporter Bioassay, Core Kit 5X, 1 each
Catalog No. PRG9995 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRG9995 1 Each
1 options
Catalog No. PRG9995 Supplier Promega Supplier No. G9995
Add to Cart
Add to Cart
Request a Quote
The FcgammaRIIa-H ADCP Reporter Bioassay is a biologically relevant MOA-based assay that can be used to measure the potency and stability of antibodies and other biologics that specifically bind and activate FcgammaRIIa.
  • Measure Potency and Stability of Biologics that Bind and Activate ADCP through FcγRIIa−H
  • Available in two kit formats:
  • Complete, everything you need to get started
  • Core, used with customer−defined Ab and target cells
Specifications
Content And Storage Components at different temps
Quantity 1 Each