Learn More
ACROBiosystems Recombinant Protein;Human Fc gamma RIIIA / CD16a (V176) Protein (SPR verified);HEK293;1MG;CD8,FCGR3A,CD16A,FCG3,FCGR3,IGFR3,FcRIIIa,FcRIII,Fc-gamma RIII,Fc-gamma RIIIa,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems CD8H52H41MG
Encompass_Preferred
Recombinant Protein;Human Fc gamma RIIIA / CD16a (V176) Protein (SPR verified);1MG;Human CD16a (V176) (SPR verified) (CD8-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.7 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16a (V176) (Cat. No. CD8-H52H4) on CM5 Chip via anti-His antibody, can bind  Herceptin with an affinity constant of 163 nM as determined in SPR assay (Biacore T200) (QC tested).
Catalog No. 50-165-8902
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.