Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR1A Mouse anti-Human, Clone: UMAB74, lyophilized, UltraMAB™
SDP

Catalog No. 501684954
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4954 Supplier OriGene Supplier No. UM500062CF
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.

Specifications

Antigen FCGR1A
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone UMAB74
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative
Gene FCGR1A
Gene Accession No. P12314
Gene Alias CD64, CD64A, FCRI, IGFR1
Gene Symbols FCGR1A
Host Species Mouse
Immunogen Full length human recombit protein of human FCGR1A produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2209
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.