Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR1A Mouse anti-Human, Unconjugated, Abnova™

Catalog No. 89339259 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-259 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-259 Supplier Abnova Corporation Supplier No. H00002209AP41
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody.

FCGR1A Monoclonal antibody specifically detects FCGR1A in Human samples. It is validated for EPair-Re.

Specifications

Antigen FCGR1A
Applications ELISA
Classification Monoclonal
Conjugate Unconjugated
Description Fc fragment of IgG, high affinity Ia, receptor (CD64)
Gene FCGR1A
Gene Alias CD64, CD64A, FCRI, FLJ18345, IGFR1
Host Species Mouse
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 2209
Includes Capture antibody: mouse monoclonal anti-FCGR1A, IgG2a Kappa (100 μg); Detection antibody: biotinylated mouse monoclonal anti-FCGR1A, IgG2a Kappa (50 μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.