Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR2A Mouse anti-Human, Clone: UMAB60, lyophilized, UltraMAB™
SDP

Catalog No. 501684936
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4936 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4936 Supplier OriGene Supplier No. UM500051CF
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.

Specifications

Antigen FCGR2A
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone UMAB60
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative
Gene FCGR2A
Gene Accession No. P12318
Gene Alias CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2
Gene Symbols FCGR2A
Host Species Mouse
Immunogen Full length human recombit protein of human FCGR2A produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2212
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.