Learn More
Invitrogen™ FCRLA (FREB) Monoclonal Antibody (N28.1), eBioscience™

Description
Description: The N28.1 monoclonal antibody recognizes the Fc receptor homolog expressed in B cells (FREB), also know as FCRLA or FcRX. FREB is a member of the family of Fc receptors for IgG (FcRg). Unlike FcRg, FREB lacks a transmembrane sequence and has an intracellular localization. FREB has been detected in proliferating and activated B cells, EBV-transformed B cells, germinal center centroblast, follicular lymphoma and the Burkitt's lymphoma cell line Raji. In contrast with published reports, the N28.1 antibody was also able to detect expression of FREB in resting B cells from normal human blood samples. Applications Reported: This N28.1 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, and immunohistochemical staining. (Fluorochrome conjugated N28.1 is recommended for use in intracellular flow cytometry). Applications Tested: This N28.1 antibody has been tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.03 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. IC Fixation Buffer (Product # 00-8222-49) and Foxp3/Transcription Factor Staining Buffer (Pr...
Specifications
Specifications
| Antigen | FCRLA (FREB) |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | N28.1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | FCRLA |
| Gene Accession No. | Q7L513 |
| Gene Alias | Fc receptor homolog expressed in B cells (FREB); fc receptor homolog expressed in B-cells; Fc receptor like A; Fc receptor related protein X; Fc receptor-like A; Fc receptor-like and mucin-like 1; fc receptor-like and mucin-like protein 1; Fc receptor-like protein; fc receptor-related protein X; FCRL; FCRL1; FCRLA; FCRLb; FCRLc1; FCRLc2; FCRLd; FCRLe; FCRLM1; FCRLX; FCRX; FREB; UNQ291/PRO329 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.