Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRLA (FREB) Monoclonal Antibody (N28.1), eBioscience™
GREENER_CHOICE

Catalog No. p-7084872
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-132-08 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-132-08 Supplier Invitrogen™ Supplier No. 14584782
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The N28.1 monoclonal antibody recognizes the Fc receptor homolog expressed in B cells (FREB), also know as FCRLA or FcRX. FREB is a member of the family of Fc receptors for IgG (FcRg). Unlike FcRg, FREB lacks a transmembrane sequence and has an intracellular localization. FREB has been detected in proliferating and activated B cells, EBV-transformed B cells, germinal center centroblast, follicular lymphoma and the Burkitt's lymphoma cell line Raji. In contrast with published reports, the N28.1 antibody was also able to detect expression of FREB in resting B cells from normal human blood samples. Applications Reported: This N28.1 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, and immunohistochemical staining. (Fluorochrome conjugated N28.1 is recommended for use in intracellular flow cytometry). Applications Tested: This N28.1 antibody has been tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.03 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. IC Fixation Buffer (Product # 00-8222-49) and Foxp3/Transcription Factor Staining Buffer (Pr...

Receptors for the Fc fragment of IgG, or FCGRs (see MIM 146790), are cell surface glycoproteins of the Ig superfamily (IgSF). These receptors mediate phagocytosis of IgG-coated pathogens and promote activation of effector cells, leading to inflammatory responses and antibody-mediated cellular cytotoxicity. All FCGR genes map to human chromosome 1. Additional genes in this region, including FREB, encode FCGR homologs that are selectively expressed in B cells and may be implicated in B-cell development and lymphomagenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRLA (FREB)
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone N28.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene FCRLA
Gene Accession No. Q7L513
Gene Alias Fc receptor homolog expressed in B cells (FREB); fc receptor homolog expressed in B-cells; Fc receptor like A; Fc receptor related protein X; Fc receptor-like A; Fc receptor-like and mucin-like 1; fc receptor-like and mucin-like protein 1; Fc receptor-like protein; fc receptor-related protein X; FCRL; FCRL1; FCRLA; FCRLb; FCRLc1; FCRLc2; FCRLd; FCRLe; FCRLM1; FCRLX; FCRX; FREB; UNQ291/PRO329
Gene Symbols FCRLA
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84824
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.