Learn More
ACROBiosystems Canine FCGRT&B2M Heterodimer Protein, His Tag&Tag Free (FCM-C52W8) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Ser 298 (FCGRT) & Val 28 - Asn 125 (B2M) (Accession # XP_038512242.1 (FCGRT) & NP_001271408.1 (B2M)).
Supplier:  ACROBiosystems FCMC52W8100UG
Encompass_Preferred
FCGRT & B2M heterodimer protein (FcRn complex) consist of two subunits: p51 (equivalent to FCGRT), and p14 (equivalent to beta-2-microglobulin), and forms an MHC class I-like heterodimer. Fc fragment of IgG, receptor, transporter, alpha (FCGRT) binds to the Fc region of monomeric immunoglobulins gamma and mediates the uptake of IgG from milk. FCGRT possible role in transfer of immunoglobulin G from mother to fetus. Beta-2-microglobulin (B2M) is a component of the class I major histocompatibility complex (MHC) and involved in the presentation of peptide antigens to the immune system.
Catalog No. 50-225-3283