Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FDC Monoclonal Antibody (CNA.42), eBioscience™, Invitrogen™

Catalog No. 5013937
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-139-37 25 μg
50-139-38 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-139-37 Supplier Invitrogen™ Supplier No. 14996880
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody CNA.42 recognizes a unique carbohydrate epitope found on follicular dendritic cells (FDCs) from a variety of species. FDCs are large cells in germinal centers (GCs) of primary and lymphoid tissues. They present non-processed antigen and switch off apoptosis of B cells through a selection process resulting in selection of memory B cells. FDCs express many B cell markers such as CD21 (long isoform), CD23 and CD35. A role for FDCs as been shown in various infectious diseases such as AIDS and transmissible spongiform encephalitis. Staining with this CNA.42 monoclonal antibody is mainly restricted to the plasma membrane but in limited cases may also be cytoplasmic (as seen in mast cells). This monoclonal has been shown to be a useful reagent for both flow cytometry and IHC analysis. Applications Reported: This CNA.42 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemistry on frozen and formalin-fixed paraffin embedded tissue sections. Applications Tested: This CNA.42 antibody has been tested by flow cytometric of a human cell line and western blotting. This can be used at less than or equal to 0.5 μg per million cells in a 100 μL total staining volume or 2 μg/mL (for western blotting). The antibody has also been tested by immunohistochemistry on formalin-fixed paraffin embedded human tonsil using high pH antigen antigen retrieval.

FDCs are large cells in germinal centers (GCs) of primary and lymphoid tissues. They present non-processed antigen and switch off apoptosis of B cells through a selection process resulting in selection of memory B cells. FDCs express many B cell markers such as CD21 (long isoform), CD23 and CD35. A role for FDCs as been shown in various infectious diseases such as AIDS and transmissible spongiform encephalitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FDC
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CNA.42
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias Dendritic Cells; FDC
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.