Learn More
MilliporeSigma™ Supelco™ Feed Tube Adapters
Catalog No. 111010824 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
25 mm
Catalog No. Description
11-101-0824 25 mm
1 options
Catalog No. 11-101-0824 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2179561EA
Ace-Thred No. 7 at top with nylon bushing and FETFE O-ring for use with feed tubes listed. Size refers to I.D. of tubing above lower thread and corresponds to M-M column diameter
TRUSTED_SUSTAINABILITY
Specifications
Description 25 mm
For Use With Feed Tubes