Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fehling's Solution 'B', Alkaline No. 2, Certified, LabChem™

Catalog No. LC142105
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC142105 20 L Cubitainer™
LC142101 500 mL Poly Bottle
LC142102 1 L Poly Bottle
LC142104 4 L Poly Bottle
LC142106 10 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC142105 Supplier LabChem, Inc. Supplier No. LC142105
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Fehling's Solution 'B', Alkaline No. 2
Name Note Alkaline No. 2
CAS Max % 60.9, 34.6, 4.5
CAS 6381-59-5, 1310-73-2
Linear Formula KNaC4H4O6·4H2O
UN Number UN1824
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 282.22
Grade Certified
Packaging Cubitainer™
Sensitivity Passes Test
Decomposition Information Sodium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Color Colorless
Quantity 20 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.