Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fehling’s Solution B Alkaline Tartrate, Reagents

Fehling’s Solution B Alkaline Tartrate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc C7202500500A

 View more versions of this product

Catalog No. 01-336-489


Add to Cart

Description

Description

Specifications

Specifications

Fehling’s Solution B Alkaline Tartrate
7732-18-5 , 6381-59-5 , 6381-59-5
Reagent
1.25
Non-sterile
Colorless
101°C
Poly Bottle
500 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.