Learn More
Fehling’s Solution B Alkaline Tartrate, Reagents
Fehling’s Solution B Alkaline Tartrate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$55.00 - $98.00
Specifications
| Chemical Name or Material | Fehling’s Solution B Alkaline Tartrate |
|---|---|
| Color | Colorless |
| CAS | 7732-18-5 , 6381-59-5 , 6381-59-5 |
| Boiling Point | 101°C |
| Grade | Reagent |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-336-492
|
Reagents Holdings Llc
C72025001A |
1 L |
Each for $98.00
|
|
|||||
|
01-336-489
|
Reagents Holdings Llc
C7202500500A |
500 mL |
Each for $55.00
|
|
|||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.