Learn More
Kingfisher Biotech Inc Feline CXCL9 (MIG) Recombinant Protein
Supplier: Kingfisher Biotech Inc RP2078F005
The Feline CXCL9 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline CXCL9 applications are for cell culture, ELISA standard, and Western Blot Control. Feline CXCL9 yeast-derived recombinant protein can be purchased in multiple sizes. Feline CXCL9 Specifications: (Molecular Weight: 12.0 kDa) (Amino Acid Sequence: TLIMRNRRCF CINTSQGTIH LKSLKDLKQF APSPSCEKTE IIVTMKNGDQ TCLNPDSADV QELIKEWEKQ VTKRKSKKKG KKNQKSRKIF KVKKSQHPHQ KKTT (104)) (Gene ID: 101100388). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.