Learn More
Raybiotech Inc Feline IFN-gamma ELISA Kit
Supplier:  Raybiotech Inc ELFIFNG1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.24 ng/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: feline Fas, IL-1 beta, IL-2, IL-4, IL-5, IL-8, IL-10, IL-12p40, RANTES
Catalog No. 50-194-8916
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.