Learn More
Kingfisher Biotech Inc Feline IL-12 p40 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2079F005
Encompass
The Feline IL-12/IL-23 p40 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline IL-12/IL-23 p40 applications are for cell culture, ELISA standard, and Western Blot Control. Feline IL-12/IL-23 p40 yeast-derived recombinant protein can be purchased in multiple sizes. Feline IL-12/IL-23 p40 Specifications: (Molecular Weight: 35.0 kDa) (Amino Acid Sequence: IWELEKNVYV VELDWHPDAP GEMVVLTCNT PEEDDITWTS DQSSEVLGSG KTLTIQVKEF ADAGQYTCHK GGEVLSHSFL LIHKKEDGIW STDILREQKE SKNKIFLKCE AKNYSGRFTC WWLTAISTDL KFTVKSSRGS SDPQEVTCGA ATLSAEKVRV DNRDYKKYTV ECQEGSACPA AEESLPIEVV VDAIHKLKYE NYTSSFFIRD IIKPDPPKNL QLKPLKNSRH VEVSWEYPDT WSTPHSYFSL TFGVQVQGKN NREKKDRLSV DKTSAKVVCH KDAKIRVQAR DRYYSSSWSN WASVSCS (307)) (Gene ID: 768273). For research use only. Made in the USA
Catalog No. 50-253-2469
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.