Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Feline IL-12p40 ELISA Kit

Catalog No. EF5RB
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Feline IL-12p40 quantitates feline IL-12p40 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant feline IL-12p40.

IL12B (IL-12 p40) encodes a subunit of interleukin 12, a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. Interleukin 12 is a disulfide-linked heterodimer composed of the 40 kDa cytokine receptor like subunit encoded by this gene, and a 35 kDa subunit encoded by IL12A. This cytokine is expressed by activated macrophages that serve as an essential inducer of Th1 cells development. This cytokine has been found to be important for sustaining a sufficient number of memory/effector Th1 cells to mediate long-term protection to an intracellular pathogen. Overexpression of this gene was observed in the central nervous system of patients with multiple sclerosis (MS), suggesting a role of this cytokine in the pathogenesis of the disease. The promoter polymorphism of this gene has been reported to be associated with the severity of atopic and non-atopic asthma in children.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CLMF; CLMF p35; CLMF p40; CLMF2; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; Il12; IL-12 p40 subunit; IL-12 subunit p35; IL-12 subunit p40; IL12, subunit p40; IL-12.p40; IL12A; IL-12 A; Il12b; IL-12 B; Il12p40; Il-12p40; ILN; IMD28; IMD29; Interleukin; Interleukin 12; interleukin 12 p40 subunit; interleukin 12, p40; interleukin 12 B; interleukin 12 B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); Interleukin12; interleukin-12 beta chain; interleukin-12 p40; interleukin-12 p40 chain; interleukin-12 p40 subunit; interleukin-12 subunit beta; interleukin-12p40; interleukin-23 p40 subunit; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p40; RP23-388G23.1
Assay Range 0.16 to 40 ng/mL
Assay Sensitivity 0.16 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 768273
Gene Symbol Il12b
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL-12 p40
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.