Learn More
Sino Biological Feline IL1B PAb 100UL 67002-T16-100
Supplier:  Sino Biological 67002T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Feline IL-1 beta (Catalog#67002-UNAE; NP_001070882.1; Ala116-Ser267). IL-1 beta specific IgG was purified by Feline IL-1 beta affinity chromatography.
Catalog No. 50-241-0323
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.