Learn More
Invitrogen™ Feline Leukemia Virus gp70 Monoclonal Antibody (C11D8)
Shop All Thermo Scientific Products

Description
MA1-82309 recognizes the gp85/gp70 envelope glycoprotein of feline leukemia virus. The antibody specifically recognizes the epitope defined by amino acids 214-218 of the gp70 sequence (aa MGPNL). Some weak cross-reactivity with the sequence MGPLL found in the FeLV p15e membrane protein may also occur. This antibody effectively neutralizes all three FeLV subgroups (A, B, and C). Removal of sodium azide is recommended prior to use in functional assays.
Specifications
Specifications
| Antigen | Feline Leukemia Virus gp70 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | C11D8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide; pH 7.2 |
| Gene Alias | FLV; FLV gp70; gp70 peptide |
| Host Species | Mouse |
| Immunogen | Proprietary Immunogen. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.