Learn More
Med Vet International Nobivac Merck, Feline Vaccine 1-HCP, 25x1 Dose Tray, Cold Ship
Supplier:  Med Vet International PHV1HCP
Encompass_Preferred
A quality 1-year core vaccine that protects against feline rhinotracheitis, feline panleukopenia virus, and feline calicivirus. Protects cats against the most common threatening viral infections calicivirus, herpes virus, and the deadly feline parvovirus, panleukopenia Nobivac Feline 1-HCP has been shown to block the replication of canine parvovirus (CPV) in cats. Non-adjuvanted formula Optimal choice for healthy indoor cats Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. *Next day air available with additional charges*
Catalog No. 50-223-2572
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Med Vet International Nobivac Merck, Feline Vaccine 1-HCP, 25x1 Dose Tray, Cold Ship
Your feedback has been submitted: Thank you for helping us improve our website.