Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEM1C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501729167 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9167 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9167 Supplier Proteintech Group Inc Supplier No. 140371AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody specifically recognizes the 69 kDa FEM1C protein.

Probable component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit.

Specifications

Antigen FEM1C
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FEM1C
Gene Accession No. Q8CEF1, Q96JP0
Gene Alias EUROIMAGE686608, EUROIMAGE783647, fem 1 homolog c (C. elegans), FEM1 gamma, FEM1A, FEM1C, KIAA1785, Protein fem 1 homolog C
Gene Symbols FEM1C
Host Species Rabbit
Immunogen FEM1C Fusion Protein Ag5099
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240263, 56929
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.