Learn More
International Medical Industries (IMI) Guarded Luer Connector (Female Luer Slip - Female Luer Lock) (Box of 50)
Supplier:  International Medical Industries (IMI) 57402
Encompass
Prep-Fill™ Guarded Luer Connector (Female Luer Slip - Female Luer Lock) provide convenient, economical, high-quality solutions to facilitate transferring or mixing of fluids between drug delivery devices or IV sets. Prep-Fill™ Guarded Luer Connectors introduce feature enhancements to essential pharmaceutical compounding devices such as expanded wing size for secure manipulation, notched wing design that enhances first air over the connection port, and built-in drip tray design to capture stray fluid droplets during repeater pump procedures. •Packaged Sterile • Non-Pyrogenic • Individual Blister • Box of 50 • Blue • Female Luer Slip - Female Luer Lock
Catalog No. NC2030206
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.