Learn More
G-Biosciences™ femtoELISA™ HRP Substrate Only
SCI_ED
Catalog No. 501036078
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
50-103-6078 100 mL
1 options
Catalog No. 50-103-6078 Supplier G-Biosciences™ Supplier No. 786111
Add to Cart
Add to Cart
femtoELISA™ HRP Substrate for highly sensitive colorimetric detection of HRP or AP.
femtoELISA™ HRP Substrate is a highly sensitive chromogen for immunoassays of horseradish peroxidase (HRP) and is based on 3,3′,5,5′‐tetramethylbenzidine (TMB). This single component substrate is ready to use and is stable for up to 2 years, if stored and used as recommended.
Specifications
Quantity 100 mL
Detection Method Colorimetric