Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fenclofenac 25mg | 34645-84-6 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY11480625MG

Encompass_Preferred

Fenclofenac is a nonsteroidal anti-inflammatory drug (NSAID) supplied as a research-grade analytical standard for inflammation studies. It exhibits antipyretic and analgesic activity and is provided as a white to off-white solid.

  • High purity (99.1%).
  • Molecular weight 297.13 g/mol.
  • Chemical formula C14H10Cl2O3.
  • White to off-white solid for analytical use.
  • Packaged as 25 MG vials, multiple sizes available.
  • Store sealed, away from moisture; follow recommended cold storage for solutions.

Catalog No. 50-002-28289


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.