Learn More
Apexbio Technology LLC Fendiline (hydrochloride) 13636-18-5 5g
SDP
Supplier:  Apexbio Technology LLC C56755000
Encompass_Preferred
Fendiline hydrochloride (CAS 13636-18-5) is a small molecule that functions as both an L-type calcium channel blocker and an antagonist of the 2-adrenergic receptor It demonstrates inhibition of L-type Ca2 currents in cardiac myocytes with an IC50 of 17 M and binds the 2-adrenergic receptor with a Kd of 2 6 M Fendiline disrupts K-Ras nanoclustering and impedes K-Ras localization to the plasma membrane (IC50 9 64 M) attenuating downstream Ras signaling and proliferation in cancer cell lines harboring oncogenic Ras mutations It has also been clinically investigated as an anti-anginal agent Fendiline is employed in research exploring calcium signaling adrenergic pathways K-Ras-dependent oncogenesis and cardiovascular pharmacology
Catalog No. 50-000-88969
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.