Learn More
Selleck Chemical LLC TMB-PS S5283-25mg
Supplier: Selleck Chemical LLC S528325MG
TMB-PS (TMBZ-PS TetraMethyl BisPhenol S Bis-(2 6dimethyl-4-hydroxyphenyl)-sulfone) is a chromogenic reagent for the detection of hydrogen peroxide e g in the enzymatic analysis of body fluids
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.