Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fennel Oil 80.0+%, TCI America™
SDP

Catalog No. F000325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F000325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F000325ML Supplier TCI America Supplier No. F000325ML

Chemical Identifiers

CAS 8006-84-6
MDL Number MFCD00146918

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Fennel Oil
RTECSNumber LJ2550000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.