Learn More
Abcam Fenofibrate, PPAR-Alpha Agonist, Solid, >99% Purity, 5 g
Description
- Molecular weight: 360.84 g/mol
- Purity: >99%
- Potency: Potent and selective PPAR-α agonist (EC50 values are 18 μM at mouse receptors and 30 μM at human receptors)
- Selectivity: 10-fold lower affinity at PPAR-γ (EC50 values are 250 μM at mouse receptors and 300 μM at human receptors)
- Function: Hypolipidemic agent
- Form: Solid
- Composition: 100% Fenofibrate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | PPAR-α |
| Shipping Condition | Ambient |
| Reactivity | Mouse, Human |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% Fenofibrate |
| Form | Solid |
| Cross Reactivity | Mouse, Human |
| Formulation | 100% Fenofibrate |
| Quantity | 5 g |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.