Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenofibrate 98.0+%, TCI America™
SDP

Catalog No. F0674100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0674100G 100 g
F06745G 5 g
F067425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F0674100G Supplier TCI America Supplier No. F0674100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49562-28-9
Molecular Formula C20H21ClO4
Molecular Weight (g/mol) 360.834
MDL Number MFCD00133314
InChI Key YMTINGFKWWXKFG-UHFFFAOYSA-N
Synonym fenofibrate, procetofen, antara, lipanthyl, lipantil, tricor, fenobrate, lipidil, secalip, fenoglide
PubChem CID 3339
ChEBI CHEBI:5001
IUPAC Name propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 81°C
Color White
Quantity 100 g
Formula Weight 360.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Fenofibrate
RTECSNumber UA2453400
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.