Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenofibrate, Tocris Bioscience™
SDP

Catalog No. 411350
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-135-0 Supplier R&D Systems Supplier No. 4113/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PPARα agonist

PPARα agonist; hypolipidemic. Attenuates insulin resistance and increases high-density lipoprotein-cholesterol in db/db mice.

Chemical Identifiers

CAS 49562-28-9
Molecular Formula C20H21ClO4
Molecular Weight (g/mol) 360.834
InChI Key YMTINGFKWWXKFG-UHFFFAOYSA-N
Synonym fenofibrate, procetofen, antara, lipanthyl, lipantil, tricor, fenobrate, lipidil, secalip, fenoglide
PubChem CID 3339
ChEBI CHEBI:5001
IUPAC Name propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl

Specifications

Quantity 50 mg
Formula Weight 360.83
Percent Purity >99%
Chemical Name or Material Fenofibrate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.