Learn More
BTNX Inc Rapid Response™ Fentanyl Test Strip (Urine) - 100 Tests
Supplier:  BTNX Inc 633640
Encompass_Preferred
The Rapid Response™ Fentanyl Test Strip (Urine) detects the presence of fentanyl in human urine specimens at a cut-off of 20 ng/mL. With an accuracy of over 99.9%, this drug test provides results in just 5 minutes. For forensic use only. Kit contains 100 Tests per kit.WARNING: This test does not evaluate drug safety or purity.It is specific for fentanyl screening with no significant cross-reactivity to other opiates, such as morphine and heroin. This kit includes 100 individually packaged test strips sealed in a pouch to prevent contamination, results interpretation printed on the back of each pouch to aid in reading the results, and instructions. The Rapid Response™ Fentanyl Test Strip is also available for Liquid/Powder (Cat. # FYL-18S7-100).
Catalog No. NC2010932