Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AarI (2 U/μL)
GSA_VA

Catalog No. p-4521484
Change view
Click to view available options
Quantity:
125 units
25 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERER1581 25 units
FERER1582 125 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERER1581 Supplier Thermo Scientific™ Supplier No. ER1581
Only null left
Add to Cart
Edge
Add to Cart

Cut at CACCTGC(4/8)^ sites with AarI restriction enzyme, which performs best at 37°C in its own unique (+oligo) buffer.

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C A C C T G C (N)4▵...3'

3'...G T G G A C G (N)8▵...5'

Conditions for 100% Activity:

  • [1X Buffer AarI] + oligonucleotide:
  • [10mM Bis-Tris Propane-HCl (pH 6.5 at 37°C), 10mM MgCl2, 100mM KCl, 0.1mg/mL BSA] + 0.5μM of oligonucleotide (see Note)
  • Incubate at 37°C

Storage Buffer:

  • AarI is supplied in:
  • 10mM potassium phosphate (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 10-fold overdigestion with AarI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — cleavage impaired
  • EcoKI: never overlaps — no effect
  • EcoBI: may overlap — effect not determined

Digestion of Agarose-embedded DNA:

  • Minimum 5 units of the enzyme are required for digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Note:

For cleavage with AarI at least two copies of its recognition sequence are required. Inclusion of 0.5μM oligonucleotide with the AarI recognition sequence in the reaction mixture significantly improves cleavage of DNAs, especially of those with a single AarI site. Still, a complete cleavage of some substrates with AarI is difficult to achieve. Greater than 10-fold overdigestion with AarI may result in star activity. AarI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme AarI
Compatible Buffer Unique Buffer (10X Buffer AarI)
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 25 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.