Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ dm4CTP

Catalog No. FERR0421 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 μmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0421 1 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0421 Supplier Thermo Scientific™ Supplier No. R0421

missing translation for 'validMainframeMsg'

Synthesize methylated DNA with the modified nucleotide dm4CTP.

dm4CTP (2'-deoxy-N4-methylcytidine 5'-triphosphate) modified nucleotide is supplied as 10 mM aqueous solution titrated to pH 7.0 with NaOH.

dm4CTP can be enzymatically incorporated into DNA with Taq DNA Polymerase, Klenow Fragment, and it is used in the synthesis of methylated DNA.

General Characteristics:
  • lambdamax = 274nm, extinction coefficient = 13.6x103 M-1 x cm-1 (pH7.0)
Quality Control:
  • Functionally tested in PCR with taq DNA Polymerase
  • Greater than 96% purity confirmed by HPLC
  • Free of endo-, exodeoxyribonuclease, phosphatase and nicking activities

Recommended for:

Synthesis of methylated DNA.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mM
Content And Storage -20°C
Quantity 1 μmol
Product Type dm4CTP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.