Learn More
Description
To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.
restriction enzymes exhibit 100% of their certified activity in the recommended buffer.
The following enzymes require unique buffers for optimal digestion:
- AarI
- AjiI
- BamHI
- BfuI
- Bpu10I
- BseXI
- Bsp143I
- Cfr9I
- Cfr10I
- Eam1105I
- Ecl136II
- Eco52I
- EcoRI
- KpnI
- Lsp1109I
- PacI
- PasI
- Ppu21I
- SacI
- ScaI
- SdaI
- SduI
- SgeI
- TaqI
Specifications
Specifications
| Concentration | 10X |
| Quantity | 1 mL |
| Product Type | Digestion Buffer |
| Research Category | Traditional Cloning |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.