Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Eco52I Buffer (10X)

Catalog No. FERB22 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERB22 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERB22 Supplier Thermo Scientific™ Supplier No. B22
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimize digestions with Eco52I restriction enzyme using this buffer, premixed with BSA for enhanced stability.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.

restriction enzymes exhibit 100% of their certified activity in the recommended buffer.

The following enzymes require unique buffers for optimal digestion:

  • AarI
  • AjiI
  • BamHI
  • BfuI
  • Bpu10I
  • BseXI
  • Bsp143I
  • Cfr9I
  • Cfr10I
  • Eam1105I
  • Ecl136II
  • Eco52I
  • EcoRI
  • KpnI
  • Lsp1109I
  • PacI
  • PasI
  • Ppu21I
  • SacI
  • ScaI
  • SdaI
  • SduI
  • SgeI
  • TaqI
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Quantity 1 mL
Product Type Eco52I Buffer
Research Category Traditional Cloning
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.