Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FastDigest TasI

Catalog No. FERFD1354 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL (100 Reactions)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERFD1354 100 μL (100 Reactions)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERFD1354 Supplier Thermo Scientific™ Supplier No. FD1354
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.

pBR322 DNA, 1.0% agarose, 8 cleavage sites

FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment (Cat. No. EP0051), T4 DNA Ligase (Cat. No. EL0014), alkaline phosphatases and T4 DNA Polymerase (Cat. No. EP0061) all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features
• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available

Applications
• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis

Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.

For methylation sensitivity, refer to product specifications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • FastDigest TasI (100 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.

Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme TasI
Compatible Buffer 10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat Inactivation No
Optimal Reaction Temperature 65°C
Type IIS RE No
Quantity 100 μL (100 Reactions)
Product Type Restriction Enzyme
Research Category Traditional Cloning
Product Line FastDigest
Overhang 5' protruding ends
Show More Show Less
Can I store the FastDigest Buffers at 4 degrees C?

The FastDigest Buffers can be stored at 4 degrees C for at least 6 months. For long-term storage until their expiration, we recommend storing the FastDigest and FastDigest Green Buffers at -20 degrees C.

How can I perform large scale digestion with the FastDigest restriction enzymes?

Our general recommendation is to use 5-10 µL of FastDigest enzymes with 100 µg DNA in 500 µL and 10-20 µL of the FastDigest enzymes with 200 µg DNA in 1 mL. Incubate the reaction for 16 h (or the longest recommended incubation time if the particular enzyme is prone to star activity) at the recommended temperature. Optimal conditions may vary as complete digestion of DNA depends on the nature of the particular FastDigest restriction enzymes and the DNA substrates.

How I can transform or calculate reaction numbers in the unit of FastDigest enzymes?

The concentration of FastDigest enzymes is proprietary information. The FastDigest enzyme activity is measured by FastDigest unit (FDU): 1 µL of enzyme (1 FDU) cleaves 1 µg of DNA substrate in 5-15 min at 37 degrees C in 20 µL of 1X FastDigest buffer.

Are the FastDigest and FastDigest Green Buffer available separately?

Yes, the FastDigest Buffer (Cat. No. B64) and the FastDigest Green Buffer (Cat. No. B71) can be purchased separately.

What is the difference between FastDigest Buffer and FastDigest Green Buffer?

The FastDigest Green Buffer offers the same performance as the colorless FastDigest Buffer but enables direct loading of the reaction mixture on gels. The 10X FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading. The blue dye migrates with 3-5 kb DNA fragments in a 1% agarose gel and has an excitation peak of 424 nm. The yellow dye migrates faster than 10 bp DNA fragments in a 1% agarose gel and has an excitation peak of 615 nm.

What is the composition of the FastDigest and FastDigest Green Buffer?

The FastDigest and FastDigest Green Buffers are proprietary digestion buffers which support 100% activity of all FastDigest restriction enzymes.

What is the unit or activity of FastDigest Restriction Enzymes?

Activity of FastDigest enzymes is defined as 1 µL of FastDigest enzyme cleaves1 µg of substrate DNA in 5 to 15 minutes in 20 µL of FastDigest buffer. This is in contrast to activity of conventional restriction enzymes which is defined as 1 unit of enzyme hydrolyzes 1 µg of lambda DNA in 60 minutes in 50 µL of optimal buffer for an enzyme.

What is the concentration of Mg2+ in the FastDigest buffers?

The composition of the FastDigest buffers is proprietary.

Is BSA present in the buffers of Thermo Scientific restriction enzymes?

As an added convenience, BSA is included in the conventional restriction enzyme buffers, as well as the FastDigest universal buffer. This eliminates the need to add BSA in a separate step.

Do FastDigest restriction enzymes contain BSA?

Yes, FastDigest restriction enzymes contain BSA.

Can I double digest my DNA using a Thermo Scientific conventional restriction enzyme and a FastDigest restriction enzyme?

For optimal results with fast reaction and 100% buffer compatibility, we highly recommend using FastDigest restriction enzymes in double digestion. In certain cases however, it may be possible to perform double digestion using a mix of Thermo Scientific conventional and Fastdigest restriction enzymes. For specific recommendations, please contact our technical service with detailed information about the enzymes and DNA template you plan to use.

Why do you recommend only 2 µL of 10X Reaction Buffer when digesting unpurified PCR product in a 30 µL reaction?

We recommend only 2 µl 10X Buffer in digestion of unpurified PCR products in 30 ul since salts and ions from the PCR reaction would be carried over to the digestion reaction.

What are key factors promoting star activity?

Star activty may be contributed by:

• Prolonged incubation
• High enzyme concentration
• High glycerol concentration (usually 5% or higher)
• Small reaction volume

Unexpected DNA bands were observed on agarose gel electrophoresis after restriction digestion. What may have caused this?

Unexpected cleavage patterns may be caused by the following reasons:

• Star activity of the restriction enzyme: Make sure to follow the reaction recommendations as specified in the protocol. Star activity may be improved by changing several key factors such as decreasing the reaction time, increasing the reaction volume, and decreasing the enzyme amount.

• Partial or incomplete cleavage (incomplete restriction reaction): Efficiency of the enzyme can be improved by adding more enzyme, prolonging the reaction time, or purifying DNA samples to remove inhibitory contaminants.

• Contamination with non-specific endonucleases: Non-specific endonucleases may be introduced to the DNA sample and/or the enzyme from improper handling, pipetting, etc.

•Improper reaction setup: Mix the digestion reaction thoroughly.

What are possible reasons for incomplete/failed restriction digestion?

The main reason for DNA cleavage reaction failure is the presence of contaminating inhibitors in the template DNA (for example: phenol, chloroform, detergents, ethanol, excess salts, EDTA, etc.). The best way to troubleshoot is to perform control reactions:

1) negative control (experimental DNA in the reaction buffer without the restriction enzyme) to access degradation of DNA by contaminants in the DNA template and/or reaction buffer
2) positive control reaction I (digestion of highly pure control DNA with the restriction enzyme) to access reaction conditions and enzyme activity
3) positive control reaction II (highly pure control DNA + experimental DNA + Restriction Enzyme) to access possible issues with the experimental DNA.

In addition, please check for sensitivity of the restriction enzymes to template DNA methylation.

Is there any other way to inactivate Thermo Scientific restriction enzymes (when thermal inactivation is not applicable) without using phenol/chloroform?

To remove the enzymes without using phenol/chloroform, we would recommend silica column based purification such as GeneJET Gel Extraction and DNA Cleanup Micro Kit (Cat. Nos. K0831/2) or GeneJET PCR Purification Kit (Cat. Nos. K0701/2).

What is the stability of Thermo Scientific restriction enzymes if they freeze-thaw during shipment?

Enzymes may freeze during shipment on dry ice. This does not affect their quality as all Thermo Scientific enzymes are tested 100% active after at least three freeze-thaw cycles. For 24-48 hour delivery, enzymes may be shipped on blue ice because their quality is not affected by short exposure to 4 degrees C.

Do Thermo Scientific restriction enzymes come with the buffer(s)?

Thermo Scientific restriction enzymes are shipped with their optimal digestion buffer(s). The buffers are available separately for purchase as well.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.