Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fluorescein-12-dUTP Solution (1 mM)

Catalog No. FERR0101 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0101 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0101 Supplier Thermo Scientific™ Supplier No. R0101
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific Fluorescein-12-dUTP (fluorescein-5(6)-carboxaminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5-triphosphate]) is supplied as 1 mM aqueous solution.

Thermo Scientific Fluorescein-12-dUTP (fluorescein-5(6)-carboxaminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5-triphosphate]) is supplied as 1 mM aqueous solution.

Fluorescein-12-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
• Taq DNA polymerase (Cat. No. EP0072)
• phi29 DNA Polymerase (EP0091)
• Klenow Fragment, exo- (EP0421)
• Klenow Fragment (EP0051)
• DNA Polymerase I (EP0041)

Applications
• Enzymatic non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random primed labeling
• Primer extension

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mM
Content And Storage Fluorescein-12-dUTP Solution, 1 mM (25 μL)

Store at –20°C.
Form Liquid
Polymerase DNA Polymerase
Labeling Method Direct Labeling
Label or Dye Fluorescein (FITC)
Quantity 25 μL
Shipping Condition Dry Ice
For Use With (Application) cDNA Synthesis, Non-Radioactive Labeling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.