Learn More
Description
Lambda DNA digested with MnlI, 1.4% agarose, 262 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...C C T C (N)7▵...3'
3'...G G A G (N)6▵...5'
Conditions for 100% Activity:
- 1X Buffer G: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 50mM NaCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- MnlI is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: may overlap —no effect
- EcoKI: never overlaps —no effect
- EcoBI: may overlap —blocked
Note:
MnlI produces DNA fragments that have a single-base 3 extension which are more difficult to ligate than blunt-ended fragments. MnlI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.
Specifications
Specifications
| Concentration | 10 U/μL |
| Methylation Sensitivity | Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive |
| Enzyme | MnlI |
| Compatible Buffer | 10X Buffer G |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | Yes |
| Quantity | 300 units |
| Product Type | Restriction Enzyme |
| Research Category | Scarless Cloning |
Frequently Asked Questions (FAQs)
For optimal results with fast reaction and 100% buffer compatibility, we highly recommend using FastDigest restriction enzymes in double digestion. In certain cases however, it may be possible to perform double digestion using a mix of Thermo Scientific conventional and Fastdigest restriction enzymes. For specific recommendations, please contact our technical service with detailed information about the enzymes and DNA template you plan to use.
We recommend only 2 µl 10X Buffer in digestion of unpurified PCR products in 30 ul since salts and ions from the PCR reaction would be carried over to the digestion reaction.
Star activty may be contributed by:
• Prolonged incubation
• High enzyme concentration
• High glycerol concentration (usually 5% or higher)
• Small reaction volume
Unexpected cleavage patterns may be caused by the following reasons:
• Star activity of the restriction enzyme: Make sure to follow the reaction recommendations as specified in the protocol. Star activity may be improved by changing several key factors such as decreasing the reaction time, increasing the reaction volume, and decreasing the enzyme amount.
• Partial or incomplete cleavage (incomplete restriction reaction): Efficiency of the enzyme can be improved by adding more enzyme, prolonging the reaction time, or purifying DNA samples to remove inhibitory contaminants.
• Contamination with non-specific endonucleases: Non-specific endonucleases may be introduced to the DNA sample and/or the enzyme from improper handling, pipetting, etc.
•Improper reaction setup: Mix the digestion reaction thoroughly.
The main reason for DNA cleavage reaction failure is the presence of contaminating inhibitors in the template DNA (for example: phenol, chloroform, detergents, ethanol, excess salts, EDTA, etc.). The best way to troubleshoot is to perform control reactions:
1) negative control (experimental DNA in the reaction buffer without the restriction enzyme) to access degradation of DNA by contaminants in the DNA template and/or reaction buffer
2) positive control reaction I (digestion of highly pure control DNA with the restriction enzyme) to access reaction conditions and enzyme activity
3) positive control reaction II (highly pure control DNA + experimental DNA + Restriction Enzyme) to access possible issues with the experimental DNA.
In addition, please check for sensitivity of the restriction enzymes to template DNA methylation.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.