Learn More
Thermo Scientific™ Rapid DNA Ligation Kit
GSA_VA
Catalog No. p-4530525
Change view
Click to view available options
Quantity:
150 Reactions
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK1422 50 Reactions
FERK1423 150 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK1422 Supplier Thermo Scientific™ Supplier No. K1422
Add to Cart
Edge
Add to Cart
Enable fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
The kit contains T4 DNA ligase and a specially formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
  • Fast—ligation of either sticky- or blunt-end DNA in only 5 minutes
  • Convenient—reaction mixture can be used directly for bacterial transformation
Recommended for:
Routine cloning experiments; Blunt-end cloning; Library construction; TA cloning.
Note:
Lyophilized bacterial strains GM2163 (†M0099) and JM107 (†M0109) can be provided free of charge upon request, with any purchase. Prior to electroporation it is necessary to inactivate T4 DNA ligase (1) by chloroform extraction.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Buffer 5X Rapid Ligation Buffer
Enzyme DNA Ligase
Product Type Rapid DNA Ligation Kit
Quantity 50 Reactions
What is the difference between Rapid DNA Ligation Kit and Thermo Scientific T4 DNA ligase with a standard buffer?
The speed of ligation is the main difference between the two. The Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature. The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation.
For Research Use Only. Not for use in diagnostic procedures.